MilliporeSigma
  • Attenuation of TRPV1 by AMG-517 after nerve injury promotes peripheral axonal regeneration in rats.
Attenuation of TRPV1 by AMG-517 after nerve injury promotes peripheral axonal regeneration in rats.
Molecular pain (2018-05-18)
Juan Bai, Fu Liu, Li-Fei Wu, Ya-Fang Wang, Xia-Qing Li
ABSTRACT
Aims The main objective was to investigate the effects of the transient receptor potential cation channel subfamily V member 1 (TRPV1) on nerve regeneration following sciatic transection injury by functional blockage of TRPV1 using AMG-517, a specific blocker of TRPV1. Methods AMG-517 was injected into the area surrounding ipsilateral lumbar dorsal root ganglia 30 min after unilateral sciatic nerve transection. The number of sciatic axons and the expression of growth-associated protein-43 (GAP-43) and glial fibrillary acidic protein was examined using semithin sections, Western blot, and immunofluorescence analyses. Results Blockage of TRPV1 with AMG-517 markedly promoted axonal regeneration, especially at two weeks after sciatic injury; the number of axons was similar to the uninjured control group. After sciatic nerve transection, expression of glial fibrillary acidic protein was decreased and GAP-43 was increased at the proximal stump. However, the expression of both glial fibrillary acidic protein and GAP-43 increased significantly in AMG-517-treated groups. Conclusions TRPV1 may be an important therapeutic target to promote peripheral nerve regeneration after injury.