MilliporeSigma
  • ISPD loss-of-function mutations disrupt dystroglycan O-mannosylation and cause Walker-Warburg syndrome.
ISPD loss-of-function mutations disrupt dystroglycan O-mannosylation and cause Walker-Warburg syndrome.
Nature genetics (2012-04-24)
Tobias Willer, Hane Lee, Mark Lommel, Takako Yoshida-Moriguchi, Daniel Beltran Valero de Bernabe, David Venzke, Sebahattin Cirak, Harry Schachter, Jiri Vajsar, Thomas Voit, Francesco Muntoni, Andrea S Loder, William B Dobyns, Thomas L Winder, Sabine Strahl, Katherine D Mathews, Stanley F Nelson, Steven A Moore, Kevin P Campbell
ABSTRACT
Walker-Warburg syndrome (WWS) is clinically defined as congenital muscular dystrophy that is accompanied by a variety of brain and eye malformations. It represents the most severe clinical phenotype in a spectrum of diseases associated with abnormal post-translational processing of a-dystroglycan that share a defect in laminin-binding glycan synthesis1. Although mutations in six genes have been identified as causes of WWS, only half of all individuals with the disease can currently be diagnosed on this basis2. A cell fusion complementation assay in fibroblasts from undiagnosed individuals with WWS was used to identify five new complementation groups. Further evaluation of one group by linkage analysis and targeted sequencing identified recessive mutations in the ISPD gene (encoding isoprenoid synthase domain containing). The pathogenicity of the identified ISPD mutations was shown by complementation of fibroblasts with wild-type ISPD. Finally, we show that recessive mutations in ISPD abolish the initial step in laminin-binding glycan synthesis by disrupting dystroglycan O-mannosylation. This establishes a new mechanism for WWS pathophysiology.
Sigma-Aldrich
Anti-Laminin α2 Antibody, clone 5H2, ascites fluid, clone 5H2, Chemicon®