MilliporeSigma
  • CAIX forms a transport metabolon with monocarboxylate transporters in human breast cancer cells.
CAIX forms a transport metabolon with monocarboxylate transporters in human breast cancer cells.
Oncogene (2019-11-15)
Samantha Ames, Jacob T Andring, Robert McKenna, Holger M Becker
ABSTRACT
Tumor cells rely on glycolysis to meet their elevated demand for energy. Thereby they produce significant amounts of lactate and protons, which are exported via monocarboxylate transporters (MCTs), supporting the formation of an acidic microenvironment. The present study demonstrates that carbonic anhydrase IX (CAIX), one of the major acid/base regulators in cancer cells, forms a protein complex with MCT1 and MCT4 in tissue samples from human breast cancer patients, but not healthy breast tissue. Formation of this transport metabolon requires binding of CAIX to the Ig1 domain of the MCT1/4 chaperon CD147 and is required for CAIX-mediated facilitation of MCT1/4 activity. Application of an antibody, directed against the CD147-Ig1 domain, displaces CAIX from the transporter and suppresses CAIX-mediated facilitation of proton-coupled lactate transport. In cancer cells, this "metabolon disruption" results in a decrease in lactate transport, reduced glycolysis, and ultimately reduced cell proliferation. Taken together, the study shows that carbonic anhydrases form transport metabolons with acid/base transporters in human tumor tissue and that these interactions can be exploited to interfere with tumor metabolism and proliferation.
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Monocarboxylate Transporter 4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GST Tag Antibody, Upstate®, from mouse
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
ExtrAvidin®-Agarose, high binding, For purification of biotinylated macromolecules and complexes
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Syrosingopine