MilliporeSigma
  • Microparticle traction force microscopy reveals subcellular force exertion patterns in immune cell-target interactions.
Microparticle traction force microscopy reveals subcellular force exertion patterns in immune cell-target interactions.
Nature communications (2020-01-09)
Daan Vorselen, Yifan Wang, Miguel M de Jesus, Pavak K Shah, Matthew J Footer, Morgan Huse, Wei Cai, Julie A Theriot
ABSTRACT
Force exertion is an integral part of cellular behavior. Traction force microscopy (TFM) has been instrumental for studying such forces, providing spatial force measurements at subcellular resolution. However, the applications of classical TFM are restricted by the typical planar geometry. Here, we develop a particle-based force sensing strategy for studying cellular interactions. We establish a straightforward batch approach for synthesizing uniform, deformable and tuneable hydrogel particles, which can also be easily derivatized. The 3D shape of such particles can be resolved with superresolution (<50 nm) accuracy using conventional confocal microscopy. We introduce a reference-free computational method allowing inference of traction forces with high sensitivity directly from the particle shape. We illustrate the potential of this approach by revealing subcellular force patterns throughout phagocytic engulfment and force dynamics in the cytotoxic T-cell immunological synapse. This strategy can readily be adapted for studying cellular forces in a wide range of applications.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)