MilliporeSigma
  • TIPE2 Induced the Proliferation, Survival, and Migration of Lung Cancer Cells Through Modulation of Akt/mTOR/NF-κB Signaling Cascade.
TIPE2 Induced the Proliferation, Survival, and Migration of Lung Cancer Cells Through Modulation of Akt/mTOR/NF-κB Signaling Cascade.
Biomolecules (2019-12-11)
Devivasha Bordoloi, Kishore Banik, Ganesan Padmavathi, Rajesh Vikkurthi, Choudhary Harsha, Nand Kishor Roy, Anuj Kumar Singh, Javadi Monisha, Hong Wang, Alan Prem Kumar, Ajaikumar B Kunnumakkara
ABSTRACT
Lung cancer represents the most common cause of cancer deaths in the world, constituting around 11.6% of all new cancer cases and 18.4% of cancer-related deaths. The propensity for early spread, lack of suitable biomarkers for early diagnosis, as well as prognosis and ineffective existing therapies, contribute to the poor survival rate of lung cancer. Therefore, there is an urgent need to develop novel biomarkers for early diagnosis and prognosis which in turn can facilitate newer therapeutic avenues for the management of this aggressive neoplasm. TIPE2 (tumor necrosis factor-α-induced protein 8-like 2), a recently identified cytoplasmic protein, possesses enormous potential in this regard. Immunohistochemical analysis showed that TIPE2 was significantly upregulated in different stages and grades of lung cancer tissues compared to normal lung tissues, implying its involvement in the positive regulation of lung cancer. Further, knockout of TIPE2 resulted in significantly reduced proliferation, survival, and migration of human lung cancer cells through modulation of the Akt/mTOR/NF-κB signaling axis. In addition, knockout of TIPE2 also caused arrest in the S phase of the cell cycle of lung cancer cells. As tobacco is the most predominant risk factor for lung cancer, we therefore evaluated the effect of TIPE2 in tobacco-mediated lung carcinogenesis as well. Our results showed that TIPE2 was involved in nicotine-, nicotine-derived nitrosamine ketone (NNK)-, N-nitrosonornicotine (NNN)-, and benzo[a]pyrene (BaP)-mediated lung cancer through inhibited proliferation, survival, and migration via modulation of nuclear factor kappa B (NF-κB)- and NF-κB-regulated gene products, which are involved in the regulation of diverse processes in lung cancer cells. Taken together, TIPE2 possesses an important role in the development and progression of lung cancer, particularly in tobacco-promoted lung cancer, and hence, specific targeting of it holds an enormous prospect in newer therapeutic interventions in lung cancer. However, these findings need to be validated in the in vivo and clinical settings to fully establish the diagnostic and prognostic importance of TIPE2 against lung cancer.
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, analytical standard
Supelco
N′-Nitrosonornicotine, analytical standard
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%