MilliporeSigma
  • Effects of the molecular weight and molar ratio of poly(2-ethyl-2-oxazoline)-based lipid on the pH sensitivity, stability, and antitumor efficacy of liposomes.
Effects of the molecular weight and molar ratio of poly(2-ethyl-2-oxazoline)-based lipid on the pH sensitivity, stability, and antitumor efficacy of liposomes.
Drug development and industrial pharmacy (2020-01-17)
Luqiao Wen, Shouzhen Huang, Weiang Du, Caili Zhu, Huan Xu
ABSTRACT
In this study, we evaluated and screened the effects of the molecular weight (MW) and molar ratio of poly(2-ethyl-2-oxazoline)-cholesteryl methyl carbonate (PEtOz-CHMC) on the pH sensitivity, stability, and antitumor efficacy of liposomes. The pH sensitivity of PEtOz-CHMC with different MWs and molar ratios was screened by drug release and cytotoxicity experiments at different pH levels. Results indicated that the liposomes coated with PEtOz1k-CHMC (7% molar ratio) and PEtOz2k-CHMC (5% molar ratio) exhibited the desirable pH responsiveness. When the MW of PEtOz was relatively low, 7% of the modified ratio obtained the strongest stability, but the turbidity of the liposomes did not obviously change when the molar ratio of PEtOz-CHMC was further increased. A375 cells were used as models to investigate the cellular uptake and intracellular localization of coumarin-6-loaded liposomes (C6-L), PEGylated liposomes (PEG-C6-L), and PEtOzylated liposomes. PEtOz1k-C6-L and PEtOz2k-C6-L presented remarkably stronger fluorescence intensity at low pH than at pH 7.4, whereas C6-L and PEG-C6-L did not achieve any obvious diversity at different pH conditions. Compared with C6-L and PEG-C6-L, PEtOz-C6-L showed efficient intracellular trafficking, including endosomal/lysosomal escape and cytoplasmic release. Pharmacokinetic experiments demonstrated that half-lives of PEG2k-C6-L, PEtOz2k-C6-L, and PEtOz1k-C6-L were 11.89-, 7.00-, and 5.29-fold times higher than those of C6-L, respectively. Among the liposomes, the DOX·HCl-loaded liposomes coated with PEtOz2k-CHMC demonstrated the strongest antitumor efficacy against B16 tumor xenograft models in vivo. These findings provide the feasibility of using PEtOz-CHMC with optimal pH sensitivity and long circulation to extend the application of liposomes to efficient anticancer drug delivery.
Sigma-Aldrich
2-Ethyl-2-oxazoline, ≥99%