MilliporeSigma
  • LncRNA MEG3 influences the proliferation and apoptosis of psoriasis epidermal cells by targeting miR-21/caspase-8.
LncRNA MEG3 influences the proliferation and apoptosis of psoriasis epidermal cells by targeting miR-21/caspase-8.
BMC molecular and cell biology (2019-10-30)
Hai-Yan Jia, Kai Zhang, Wen-Jing Lu, Gui-Wen Xu, Jian-Fen Zhang, Zhan-Li Tang
ABSTRACT
It was reported that microRNA-21(miR-21) was differentially expressed in the keratinocytes of psoriasis patients, and it may influence the apoptosis and proliferation of cells. The role of lncRNA maternally expressed gene3 (MEG3), a competing endogenous RNAs of miR-21, in the progression of psoriasis remains unclear. We aimed to unfold the influence of MEG3 and miR-21 on the proliferation and apoptosis of psoriasis epidermal cells. 50μg/L TNF-α was used to treat HaCaTs and NHEKs cells for 24 h, and then different experiments were conducted. qRT-PCR were applied for measuring the mRNA level of MEG3, miR-2, and caspase-8, and the protein expression of caspase-8 was measured with western blotting. Flow cytometry was used for assessing apoptosis. Cell proliferation was detected using MTT and colony formation assays. Dual luciferase reporter assay was applied for confirming the binding site between MEG3 and miR-21, miR-21 and Caspase-8. A cell model for in vitro studying the role of MEG3 in psoriasis pathophysiology was established using HaCaT and HHEKs. MEG3 was significantly down-regulated in HaCaT, HHEKs, and psoriatic skin samples. MEG3 inhibits proliferation and promotes apoptosis of Activated-HaCaT (Act-HaCaT) and Activated-HHEKs (Act- HHEK) by regulating miR-21, and the binding site between MEG3 and miR-21 was identified. We also found that miR-21 could inhibit the level of caspase-8 and identified the binding site between caspase-8 and miR-21. Some down-stream proteins of caspase-8, Cleaved caspase-8, cytc, and apaf-1 were regulated by miR-21 and MEG3. MEG3/miR-21 axis may regulate the expression of caspase-8, and further influence the proliferation and apoptosis of psoriasis keratinocyte, Act-HaCaT and Act- HHEK. Therefore, our findings may provide a new thought for the study of pathogenesis and treatment of psoriasis.
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human CASP8