MilliporeSigma
  • Preformulation Characterization and Stability Assessments of Secretory IgA Monoclonal Antibodies as Potential Candidates for Passive Immunization by Oral Administration.
Preformulation Characterization and Stability Assessments of Secretory IgA Monoclonal Antibodies as Potential Candidates for Passive Immunization by Oral Administration.
Journal of pharmaceutical sciences (2019-08-02)
Yue Hu, Ozan S Kumru, Jian Xiong, Lorena R Antunez, John Hickey, Yang Wang, Lisa Cavacini, Mark Klempner, Sangeeta B Joshi, David B Volkin
ABSTRACT
Enterotoxigenic Escherichia coli (ETEC) is a major cause of diarrheal disease among children in developing countries, and there are no licensed vaccines to protect against ETEC. Passive immunization by oral delivery of ETEC-specific secretory IgAs (sIgAs) could potentially provide an alternative approach for protection in targeted populations. In this study, a series of physiochemical techniques and an in vitro gastric digestion model were used to characterize and compare key structural attributes and stability profiles of 3 anti-heat-labile enterotoxin mAbs (sIgA1, sIgA2, and IgG1 produced in CHO cells). The mAbs were evaluated in terms of primary structure, N-linked glycan profiles, size and aggregate content, relative apparent solubility, conformational stability, and in vitro antigen binding. Compared to IgG1 mAb, sIgA1 and sIgA2 mAbs showed increased sample heterogeneity, especially in terms of N-glycan composition and the presence of higher molecular weight species. The sIgA mAbs showed overall better physical stability and were more resistant to loss of antigen binding activity during incubation at low pH, 37°C with pepsin. These results are discussed in terms of future challenges to design stable, low-cost formulations of sIgA mAbs as an oral supplement for passive immunization to protect against enteric diseases in the developing world.
Sigma-Aldrich
Heat-Labile Enterotoxin, B subunit (LTB) from E. coli, recombinant, expressed in Pichia pastoris, >90% (SDS-PAGE), lyophilized powder