MilliporeSigma
  • Activity of NaV1.2 promotes neurodegeneration in an animal model of multiple sclerosis.
Activity of NaV1.2 promotes neurodegeneration in an animal model of multiple sclerosis.
JCI insight (2016-11-25)
Benjamin Schattling, Walid Fazeli, Birgit Engeland, Yuanyuan Liu, Holger Lerche, Dirk Isbrandt, Manuel A Friese
ABSTRACT
Counteracting the progressive neurological disability caused by neuronal and axonal loss is the major unmet clinical need in multiple sclerosis therapy. However, the mechanisms underlying irreversible neuroaxonal degeneration in multiple sclerosis and its animal model experimental autoimmune encephalomyelitis (EAE) are not well understood. A long-standing hypothesis holds that the distribution of voltage-gated sodium channels along demyelinated axons contributes to neurodegeneration by increasing neuroaxonal sodium influx and energy demand during CNS inflammation. Here, we tested this hypothesis in vivo by inserting a human gain-of-function mutation in the mouse NaV1.2-encoding gene Scn2a that is known to increase NaV1.2-mediated persistent sodium currents. In mutant mice, CNS inflammation during EAE leads to elevated neuroaxonal degeneration and increased disability and lethality compared with wild-type littermate controls. Importantly, immune cell infiltrates were not different between mutant EAE mice and wild-type EAE mice. Thus, this study shows that increased neuronal NaV1.2 activity exacerbates inflammation-induced neurodegeneration irrespective of immune cell alterations and identifies NaV1.2 as a promising neuroprotective drug target in multiple sclerosis.
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Sodium Channel Antibody, Voltage Gated, Brain Type II, Chemicon®, from rabbit