MilliporeSigma
  • Posttranslational modifications of blood-derived alpha-synuclein as biochemical markers for Parkinson's disease.
Posttranslational modifications of blood-derived alpha-synuclein as biochemical markers for Parkinson's disease.
Scientific reports (2017-10-24)
Hugo Vicente Miranda, Rafaela Cássio, Leonor Correia-Guedes, Marcos António Gomes, Ana Chegão, Elisa Miranda, Tiago Soares, Miguel Coelho, Mário Miguel Rosa, Joaquim J Ferreira, Tiago Fleming Outeiro
ABSTRACT
Parkinson's disease (PD) is a progressive neurodegenerative disorder known for the typical motor features associated. Pathologically, it is characterized by the intracellular accumulation of alpha-synuclein (aSyn) in Lewy bodies and Lewy neurites. Currently, there are no established biochemical markers for diagnosing or for following disease progression, a major limitation for the clinical practice. Posttranslational modifications (PTMs) in aSyn have been identified and implicated on its pathobiology. Since aSyn is abundant in blood erythrocytes, we aimed to evaluate whether PTMs of aSyn in the blood might hold value as a biomarker for PD. We examined 58 patients with PD and 30 healthy age-matched individuals. We found that the levels of Y125 phosphorylated, Y39 nitrated, and glycated aSyn were increased in PD, while those of SUMO were reduced. A combinatory analysis of the levels of these PTMs resulted in an increased sensitivity, with an area under curve (AUC) of 0.843 for PD versus healthy controls, and correlated with disease severity and duration. We conclude that the levels of these selected PTMs hold strong potential as biochemical markers for PD. Ultimately, our findings might facilitate the monitoring of disease progression in clinical trials, opening the possibility for developing more effective therapies against PD.
Sigma-Aldrich
Anti-nitro-α/β-Synuclein (Tyr39) Antibody, clone nSyn14, clone nSyn14, Upstate®, from mouse