MilliporeSigma
  • Delivery of paclitaxel using nanoparticles composed of poly(ethylene oxide)-b-poly(butylene oxide) (PEO-PBO).
Delivery of paclitaxel using nanoparticles composed of poly(ethylene oxide)-b-poly(butylene oxide) (PEO-PBO).
Colloids and surfaces. B, Biointerfaces (2017-11-13)
Lijiang Wang, Ju Yao, Xiaomin Zhang, Yingxin Zhang, Chang Xu, Robert J Lee, Gary Yu, Bo Yu, Lesheng Teng
ABSTRACT
An amphiphilic block copolymer poly(ethylene oxide)-b-poly(butylene oxide) (PEO-PBO) was evaluated as a carrier for therapeutic delivery of paclitaxel (PTX). PEO-PBO and PTX form nanoparticles (NPs) by self-assembly upon hydration. The size of these NPs was about 92.71nm and the zeta potential was -5.06mV, which met the requirements for passive tumor targeting through the enhanced permeability and retention effect. Compared with a commonly used block copolymer poly(ethylene glycol)-b-poly-D,L-(lactic acid) (PEG-PDLLA), PEO-PBO forms nanoparticles with superior pharmacokinetic, biodistribution, and tumor inhibitory properties. Meanwhile, results of hemolysis study and CMC determination showed that PEO-PBO had better biocompatibility and stability than PEG-PDLLA. These data suggest that PEO-PBO has potential for application in drug delivery and warrant further evaluation.
Sigma-Aldrich
Poly(ethylene oxide)-block-poly(butylene oxide), PEG average Mn 2,000, average Mn 2,000 (PBO)
Sigma-Aldrich
Poly(ethylene oxide)-block-poly(butylene oxide), PEG average Mn 2,000, average Mn 5,000 (PBO)