MilliporeSigma
  • EPC1/TIP60-Mediated Histone Acetylation Facilitates Spermiogenesis in Mice.
EPC1/TIP60-Mediated Histone Acetylation Facilitates Spermiogenesis in Mice.
Molecular and cellular biology (2017-07-12)
Yixin Dong, Kyo-Ichi Isono, Kazuyuki Ohbo, Takaho A Endo, Osamu Ohara, Mamiko Maekawa, Yoshiro Toyama, Chizuru Ito, Kiyotaka Toshimori, Kristian Helin, Narumi Ogonuki, Kimiko Inoue, Atsuo Ogura, Kazutsune Yamagata, Issay Kitabayashi, Haruhiko Koseki
ABSTRACT
Global histone hyperacetylation is suggested to play a critical role for replacement of histones by transition proteins and protamines to compact the genome during spermiogenesis. However, the underlying mechanisms for hyperacetylation-mediated histone replacement remains poorly understood. Here, we report that EPC1 and TIP60, two critical components of the mammalian nucleosome acetyltransferase of H4 (NuA4) complexes, are coexpressed in male germ cells. Strikingly, genetic ablation of either Epc1 or Tip60 disrupts hyperacetylation and impairs histone replacement, in turn causing aberrant spermatid development. Taking these observations together, we reveal an essential role of the NuA4 complexes for histone hyperacetylation and subsequent compaction of the spermatid genome.
Sigma-Aldrich
Anti-acetyl-Histone H2B Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-TIP60 Rabbit pAb, liquid, Calbiochem®
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests