MilliporeSigma
  • Xenobiotics in vitro: the influence of L-cystine, pantothenat, and miliacin on metabolic and proliferative capacity of keratinocytes.
Xenobiotics in vitro: the influence of L-cystine, pantothenat, and miliacin on metabolic and proliferative capacity of keratinocytes.
Cutaneous and ocular toxicology (2006-05-17)
D Hoeller Obrigkeit, T Oepen, F K Jugert, H F Merk, J Kubicki
ABSTRACT
To investigate the effect of cell growth-stimulating agents on human epidermal keratinocytes, we exposed monolayers of normal human keratinocytes derived from foreskin to different concentrations of the amino acid L-cystine, the member of the vitamin B family D-pantothenat, the phytosterol miliacin, and a combination thereof in keratinocyte growth medium. As a test system for the metabolic capacity, we used the activity of mitochondrial deyhdrogenases as measured by XTT, and for the cell proliferation, we determined the BrdU-uptake. The additives, active ingredients of the hair growth drug PRIORIN, were added in the presence of fully supplemented keratinocyte growth medium or a deficient medium without L-cystine, L-methionine, L-histidin, D-pantothenat, epidermal growth factor, and bovine pituary gland extract. Deficient medium itself reduced the metabolic capacity of keratinocytes to 35% compared with keratinocytes in fully supplemented growth medium. In deficient medium cell, proliferation was not measurable. Increasing doses of L-cystine restored the reduced metabolic capacity from 46% (0.009 mg/L) to 54% (0.09 mg/L) and 92% (0.45 mg/L) in deficient medium. Addition of D-pantothenat (0.43 mg/L) enhanced the metabolic capacity to 150% only in fully supplemented growth medium, compared with untreated controls with growth medium. Miliacin (6 mg/mL) increased not only the metabolic capacity (162%) but also stimulated cell proliferation (215%) as measured by BrdU-uptake in growth medium. The combination of all three additives increased the metabolic capacity (245%) synergistically in growth medium. We were able to show effects of D-panthenol, L-lysine, and miliacin on proliferation and metabolic capacity of keratinocyte monocell culture, which was further increased by combination of the three substances. These basic results suggest a beneficial effect on keratinocyte growth and stimulation by products combining these substances (e.g., Priorin). Furthermore, this work emphasizes the suitability of keratinocyte monolayers for pharmacological testings.
Sigma-Aldrich
D-Pantothenic acid hemicalcium salt, ≥98.0%
Sigma-Aldrich
D-Panthenol, ≥98.0% (NT)