MilliporeSigma
  • Novel measurements of mammary stem cells in human umbilical cord blood as prospective predictors of breast cancer susceptibility in later life.
Novel measurements of mammary stem cells in human umbilical cord blood as prospective predictors of breast cancer susceptibility in later life.
Annals of oncology : official journal of the European Society for Medical Oncology (2011-04-26)
L Qiu, H P Low, C-I Chang, W C Strohsnitter, M Anderson, K Edmiston, H-O Adami, A Ekbom, P Hall, P Lagiou, D Trichopoulos, C-C Hsieh
ABSTRACT
The size of the breast stem-cell pool could underlie the intrauterine roots of breast cancer. We studied whether breast stem cells exist in umbilical cord blood and if they correlate with hematopoietic stem-cell measurements that have been positively associated with perinatal risk factors for breast cancer. We isolated mononuclear cells from umbilical cord blood of 170 singleton full-term pregnancies and determined, by reverse transcription polymerase chain reaction, the presence of genes of putative breast epithelial stem-cell/progenitor markers [including epithelial cell adhesion molecule (EpCAM), CD49f (α6-integrin), CD117 (c-kit receptor), CD24, and CD29 (β1-integrin)]. By immunocytochemistry, we colocalized protein expressions of EpCAM+CD49f+, CD49f+CD24+, and CD24+CD29+. We correlated concentrations of putative breast stem-cell/progenitor subpopulations, quantified by flow cytometry, with concentrations of hematopoietic stem cells. Mammary stem-cell phenotypes were identified in umbilical cord blood. The measured EpCAM+ subpopulation was positively correlated with concentrations of CD34+ and CD34+CD38- hematopoietic stem cells (both P=0.006). Additionally, EpCAM+CD49f+ and CD49f+CD24+ subpopulations were positively correlated to the CD34+ cells (P=0.03 and 0.008, respectively). The positive association between measurable breast and hematopoietic stem cells in human umbilical cord blood suggests plausible mechanisms for a prenatal influence on breast cancer risk.
Sigma-Aldrich
Anti-CD24 Antibody, clone SN3, clone SN3, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α6 Antibody, clone NKI-GoH3, clone NKI-GoH3, Chemicon®, from rat