MilliporeSigma
  • CSL controls telomere maintenance and genome stability in human dermal fibroblasts.
CSL controls telomere maintenance and genome stability in human dermal fibroblasts.
Nature communications (2019-08-31)
Giulia Bottoni, Atul Katarkar, Beatrice Tassone, Soumitra Ghosh, Andrea Clocchiatti, Sandro Goruppi, Pino Bordignon, Paris Jafari, Fabio Tordini, Thomas Lunardi, Wolfram Hoetzenecker, Victor Neel, Joachim Lingner, G Paolo Dotto
ABSTRACT
Genomic instability is a hallmark of cancer. Whether it also occurs in Cancer Associated Fibroblasts (CAFs) remains to be carefully investigated. Loss of CSL/RBP-Jκ, the effector of canonical NOTCH signaling with intrinsic transcription repressive function, causes conversion of dermal fibroblasts into CAFs. Here, we find that CSL down-modulation triggers DNA damage, telomere loss and chromosome end fusions that also occur in skin Squamous Cell Carcinoma (SCC)-associated CAFs, in which CSL is decreased. Separately from its role in transcription, we show that CSL is part of a multiprotein telomere protective complex, binding directly and with high affinity to telomeric DNA as well as to UPF1 and Ku70/Ku80 proteins and being required for their telomere association. Taken together, the findings point to a central role of CSL in telomere homeostasis with important implications for genomic instability of cancer stromal cells and beyond.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-UPF1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, Ab1