MilliporeSigma
  • Molecular characterization of autophagic and apoptotic signaling induced by sorafenib in liver cancer cells.
Molecular characterization of autophagic and apoptotic signaling induced by sorafenib in liver cancer cells.
Journal of cellular physiology (2018-08-23)
María A Rodríguez-Hernández, Raúl González, Ángel J de la Rosa, Paloma Gallego, Raquel Ordóñez, Elena Navarro-Villarán, Laura Contreras, Mario Rodríguez-Arribas, Javier González-Gallego, José M Álamo-Martínez, Luís M Marín-Gómez, José A Del Campo, José L Quiles, José M Fuentes, Jesús de la Cruz, José L Mauriz, Francisco J Padillo, Jordi Muntané
ABSTRACT
Sorafenib is the unique accepted molecular targeted drug for the treatment of patients in advanced stage of hepatocellular carcinoma. The current study evaluated cell signaling regulation of endoplasmic reticulum (ER) stress, c-Jun-N-terminal kinase (JNK), Akt, and 5'AMP-activated protein kinase (AMPK) leading to autophagy and apoptosis induced by sorafenib. Sorafenib induced early (3-12 hr) ER stress characterized by an increase of Ser51 P-eIF2α/eIF2α, C/EBP homologous protein (CHOP), IRE1α, and sXBP1, but a decrease of activating transcription factor 6 expression, overall temporally associated with the increase of Thr183,Tyr185 P-JNK1/2/JNK1/2, Thr172 P-AMPKα, Ser413 P-Foxo3a, Thr308 P-AKt/AKt and Thr32 P-Foxo3a/Foxo3a ratios, and reduction of Ser2481 P-mammalian target of rapamycin (mTOR)/mTOR and protein translation. This pattern was related to a transient increase of tBid, Bim EL , Beclin-1, Bcl-xL, Bcl-2, autophagy markers, and reduction of myeloid cell leukemia-1 (Mcl-1) expression. The progressive increase of CHOP expression, and reduction of Thr308 P-AKt/AKt and Ser473 P-AKt/AKt ratios were associated with the reduction of autophagic flux and an additional upregulation of Bim EL expression and caspase-3 activity (24 hr). Small interfering-RNA (si-RNA) assays showed that Bim, but not Bak and Bax, was involved in the induction of caspase-3 in sorafenib-treated HepG2 cells. Sorafenib increased autophagic and apoptotic markers in tumor-derived xenograft model. In conclusion, the early sorafenib-induced ER stress and regulation of JNK and AMPK-dependent signaling were related to the induction of survival autophagic process. The sustained drug treatment induced a progressive increase of ER stress and PERK-CHOP-dependent rise of Bim EL , which was associated with the shift from autophagy to apoptosis. The kinetic of Bim EL expression profile might also be related to the tight balance between AKt- and AMPK-related signaling leading to Foxo3a-dependent BIM EL upregulation.
Sigma-Aldrich
Anti-MCL1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-JNK1/2 (pThr183/pTyr185) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-FHIT (pTyr114) antibody produced in rabbit, affinity isolated antibody
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests