MilliporeSigma
  • Damage-induced lncRNAs control the DNA damage response through interaction with DDRNAs at individual double-strand breaks.
Damage-induced lncRNAs control the DNA damage response through interaction with DDRNAs at individual double-strand breaks.
Nature cell biology (2017-11-29)
Flavia Michelini, Sethuramasundaram Pitchiaya, Valerio Vitelli, Sheetal Sharma, Ubaldo Gioia, Fabio Pessina, Matteo Cabrini, Yejun Wang, Ilaria Capozzo, Fabio Iannelli, Valentina Matti, Sofia Francia, G V Shivashankar, Nils G Walter, Fabrizio d'Adda di Fagagna
ABSTRACT
The DNA damage response (DDR) preserves genomic integrity. Small non-coding RNAs termed DDRNAs are generated at DNA double-strand breaks (DSBs) and are critical for DDR activation. Here we show that active DDRNAs specifically localize to their damaged homologous genomic sites in a transcription-dependent manner. Following DNA damage, RNA polymerase II (RNAPII) binds to the MRE11-RAD50-NBS1 complex, is recruited to DSBs and synthesizes damage-induced long non-coding RNAs (dilncRNAs) from and towards DNA ends. DilncRNAs act both as DDRNA precursors and by recruiting DDRNAs through RNA-RNA pairing. Together, dilncRNAs and DDRNAs fuel DDR focus formation and associate with 53BP1. Accordingly, inhibition of RNAPII prevents DDRNA recruitment, DDR activation and DNA repair. Antisense oligonucleotides matching dilncRNAs and DDRNAs impair site-specific DDR focus formation and DNA repair. We propose that DDR signalling sites, in addition to sharing a common pool of proteins, individually host a unique set of site-specific RNAs necessary for DDR activation.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-RNF168 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Rad50 Antibody, clone 13B3/2C6, clone 13B3/2C6, Upstate®, from mouse