MilliporeSigma
  • Nucleoside analogue 2'-C-methylcytidine inhibits hepatitis E virus replication but antagonizes ribavirin.
Nucleoside analogue 2'-C-methylcytidine inhibits hepatitis E virus replication but antagonizes ribavirin.
Archives of virology (2017-06-18)
Changbo Qu, Lei Xu, Yuebang Yin, Maikel P Peppelenbosch, Qiuwei Pan, Wenshi Wang
ABSTRACT
Hepatitis E virus (HEV) infection has emerged as a global health issue, but no approved medication is available. The nucleoside analogue 2'-C-methylcytidine (2CMC), a viral polymerase inhibitor, has been shown to inhibit infection with a variety of viruses, including hepatitis C virus (HCV). Here, we report that 2CMC significantly inhibits the replication of HEV in a subgenomic replication model and in a system using a full-length infectious virus. Importantly, long-term treatment with 2CMC did not result in a loss of antiviral potency, indicating a high barrier to drug resistance development. However, the combination of 2CMC with ribavirin, an off-label treatment for HEV, exerts antagonistic effects. Our results indicate that 2CMC serves as a potential antiviral drug against HEV infection.
Sigma-Aldrich
Anti-Hepatitis E Virus Antibody, aa 434-457, clone 1E6, clone 1E6, Chemicon®, from mouse