MilliporeSigma
  • CREB overexpression in dorsal CA1 ameliorates long-term memory deficits in aged rats.
CREB overexpression in dorsal CA1 ameliorates long-term memory deficits in aged rats.
eLife (2017-01-05)
Xiao-Wen Yu, Daniel M Curlik, M Matthew Oh, Jerry Cp Yin, John F Disterhoft
ABSTRACT
The molecular mechanisms underlying age-related cognitive deficits are not yet fully elucidated. In aged animals, a decrease in the intrinsic excitability of CA1 pyramidal neurons is believed to contribute to age-related cognitive impairments. Increasing activity of the transcription factor cAMP response element-binding protein (CREB) in young adult rodents facilitates cognition, and increases intrinsic excitability. However, it has yet to be tested if increasing CREB expression also ameliorates age-related behavioral and biophysical deficits. To test this hypothesis, we virally overexpressed CREB in CA1 of dorsal hippocampus. Rats received CREB or control virus, before undergoing water maze training. CREB overexpression in aged animals ameliorated the long-term memory deficits observed in control animals. Concurrently, cells overexpressing CREB in aged animals had reduced post-burst afterhyperpolarizations, indicative of increased intrinsic excitability. These results identify CREB modulation as a potential therapy to treat age-related cognitive decline.
Sigma-Aldrich
Anti-CREB Antibody, clone NL904, rabbit monoclonal, culture supernatant, clone NL904, Upstate®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse