MilliporeSigma
  • Drebrin E is involved in the regulation of axonal growth through actin-myosin interactions.
Drebrin E is involved in the regulation of axonal growth through actin-myosin interactions.
Journal of neurochemistry (2009-02-19)
Toshiyuki Mizui, Nobuhiko Kojima, Hiroyuki Yamazaki, Michihito Katayama, Kenji Hanamura, Tomoaki Shirao
ABSTRACT
Drebrin is a well-known side-binding protein of F-actin in the brain. Immunohistochemical data suggest that the peripheral parts of growing axons are enriched in the drebrin E isoform and mature axons are not. It has also been observed that drebrin E is concentrated in the growth cones of PC12 cells. These data strongly suggest that drebrin E plays a role in axonal growth during development. In this study, we used primary hippocampal neuronal cultures to analyze the role of drebrin E. Immunocytochemistry showed that within axonal growth cones drebrin E specifically localized to the transitional zone, an area in which dense networks of F-actins and microtubules overlapped. Over-expression of drebrin E caused drebrin E and F-actin to accumulate throughout the growth cone and facilitated axonal growth. In contrast, knockdown of drebrin E reduced drebrin E and F-actin in the growth cone and prevented axonal growth. Furthermore, inhibition of myosin II ATPase masked the promoting effects of drebrin E over-expression on axonal growth. These results suggest that drebrin E plays a role in axonal growth through actin-myosin interactions in the transitional zone of axonal growth cones.
Sigma-Aldrich
Anti-Tau-1 Antibody, clone PC1C6, clone PC1C6, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Synapsin-1 Antibody, clone 10.22, clone 10.22, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid