MilliporeSigma
  • A direct interaction between the RAG2 C terminus and the core histones is required for efficient V(D)J recombination.
A direct interaction between the RAG2 C terminus and the core histones is required for efficient V(D)J recombination.
Immunity (2005-08-23)
Kelly L West, Netai C Singha, Pablo De Ioannes, Lynne Lacomis, Hediye Erdjument-Bromage, Paul Tempst, Patricia Cortes
ABSTRACT
V(D)J recombination is a tightly controlled process of somatic recombination whose regulation is mediated in part by chromatin structure. Here, we report that RAG2 binds directly to the core histone proteins. The interaction with histones is observed in developing lymphocytes and within the RAG1/RAG2 recombinase complex in a manner that is dependent on the RAG2 C terminus. Amino acids within the plant homeo domain (PHD)-like domain as well as a conserved acidic stretch of the RAG2 C terminus that is considered to be a linker region are important for this interaction. Point mutations that disrupt the RAG2-histone association inhibit the efficiency of the V(D)J recombination reaction at the endogenous immunoglobulin locus, with the most dramatic effect in the V to DJ(H) rearrangement.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H2B Antibody, Upstate®, from rabbit