MilliporeSigma
  • Identification of motifs for cell adhesion within the repeated domains of transforming growth factor-beta-induced gene, betaig-h3.
Identification of motifs for cell adhesion within the repeated domains of transforming growth factor-beta-induced gene, betaig-h3.
The Journal of biological chemistry (2000-07-25)
J E Kim, S J Kim, B H Lee, R W Park, K S Kim, I S Kim
ABSTRACT
betaig-h3 is a transforming growth factor-beta-inducible cell adhesion molecule that has four characteristic homologous repeated domains. We made recombinant betaig-h3 proteins, which were highly active in mediating human corneal epithelial (HCE) cell adhesion and spreading. The 2nd and the 4th repeated domains were sufficient to mediate HCE cell adhesion. A sequence analysis showed that aspartic acid (Asp) and isoleucine (Ile) of the 2nd and the 4th domains are highly conserved in many fasciclin 1 homologous (fas-1) domains. Substitution mutational study identified these two amino acids are essential for cell adhesion. Synthetic peptides containing Asp and Ile, NKDIL and EPDIM derived from the 2nd and the 4th domains, respectively, almost completely blocked cell adhesion mediated by not only wild type betaig-h3 but also each of the 2nd and the 4th domains. These peptides alone were fully active in mediating cell adhesion. In addition, we demonstrated the functional receptor for betaig-h3 is alpha(3)beta(1) integrin. These results, therefore, establish the essential motifs within the 2nd and the 4th domains of betaig-h3, which interact with alpha(3)beta(1) integrin to mediate HCE cell adhesion to betaig-h3 and suggest that other proteins containing Asp-Ile in their fas-1 domains could possibly function as cell adhesion molecules.
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone ASC-1 (Azide Free), clone ASC-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 12G10, clone 12G10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αV Antibody, clone P3G8, azide free, clone P3G8, Chemicon®, from mouse