MilliporeSigma
  • The ubiquitin ligase KBTBD8 regulates PKM1 levels via Erk1/2 and Aurora A to ensure oocyte quality.
The ubiquitin ligase KBTBD8 regulates PKM1 levels via Erk1/2 and Aurora A to ensure oocyte quality.
Aging (2019-02-21)
Yan-Ru Li, Rui-Rui Peng, Wen-Yi Gao, Peng Liu, Liang-Jian Chen, Xiao-Lan Zhang, Na-Na Zhang, Yang Wang, Lei Du, Feng-Yu Zhu, Li-Li Wang, Cong-Rong Li, Wen-Tao Zeng, Jian-Min Li, Fan Hu, Dong Zhang, Zhi-Xia Yang
ABSTRACT
Tight control of energy metabolism is essential for normal cell function and organism survival. PKM (pyruvate kinase, muscle) isoforms 1 and 2 originate from alternative splicing of PKM pre-mRNA. They are key enzymes in oxidative phosphorylation and aerobic glycolysis, respectively, and are essential for ATP generation. The PKM1:PKM2 expression ratio changes with development and differentiation, and may also vary under metabolic stress and other conditions. Until now, there have been no reports about the function and regulation of PKM isozymes in oocytes. Here, we demonstrate that PKM1 or PKM2 depletion significantly disrupts ATP levels and mitochondrial integrity, and exacerbates free-radical generation and apoptosis in mouse oocytes. We also show that KBTBD8, a female fertility factor in the KBTBD ubiquitin ligase family, selectively regulates PKM1 levels through a signaling cascade that includes Erk1/2 and Aurora A kinases as intermediates. Finally, using RNA sequencing and protein network analysis, we identify several regulatory proteins that may be govern generation of mature PKM1 mRNA. These results suggest KBTBD8 affects PKM1 levels in oocytes via a KBTBD8→Erk1/2→Aurora A axis, and may also affect other essential processes involved in maintaining oocyte quality.
Sigma-Aldrich
Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal, clone MAPK-YT, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-Aurora Kinase (pThr288) antibody produced in rabbit, affinity isolated antibody