MilliporeSigma
  • High-risk human papillomavirus types in HIV-infected and HIV-uninfected young women in KwaZulu-Natal, South Africa: implications for vaccination.
High-risk human papillomavirus types in HIV-infected and HIV-uninfected young women in KwaZulu-Natal, South Africa: implications for vaccination.
Infectious diseases (London, England) (2017-04-14)
Joyce N Mbatha, Myra Taylor, Elisabeth Kleppa, Kristine Lillebo, Hashini N Galappaththi-Arachchige, Deepak Singh, Eyrun F Kjetland, Marc F D Baay, Zilungile L Mkhize-Kwitshana
ABSTRACT
High-risk human papillomavirus (hr-HPV) infections and low-grade squamous intraepithelial lesions occur frequently in young women. The available vaccines cover up to seven hr-HPV genotypes (HPV16, HPV18, HPV31, HPV33, HPV45, HPV52 and HPV58) and two low-risk HPV types (HPV6 and HPV11). The objective of this study was to describe the hr-HPV genotypes present among HIV-uninfected and HIV-infected young women in rural high schools. Cervicovaginal lavages were obtained from sexually active young women recruited from high schools in KwaZulu-Natal (n = 1223). HPV testing was done by the polymerase chain reaction using GP5+/GP6 + primers and enzyme immunoassay. HIV testing was done using rapid test kits. Of the 1223 cervicovaginal lavages, 301 (25%) were positive for hr-HPV. The HPV prevalence was higher in HIV infected (32.20%, 95% CI: 0.27-0.38) than in HIV-uninfected women (22.50%, 95% CI: 0.21-0.26), (p = .001). Similarly, multiple infections were slightly more common in HIV infected (59.32%) than in HIV-uninfected women (53.51%), (p = .37). The nine predominant genotypes in descending order were HPV types 16 (n = 99, 22.10%), 51 (n = 58, 12.91%), 18 (n = 56, 12.50%), 35 (n = 50, 11.10%), 33 (n = 47, 10.82%), 56 (n = 42, 9.31%), 45 (n = 34, 7.60%), 52 (n = 32, 7.14%) and 59 (n = 31, 6.91%). HPV 35, 51, 56 and 59 (40.62%), which are not covered by any vaccine, were among the most prevalent in the schools of KwaZulu-Natal. Four of the most predominant high-risk HPV types in this region are not covered by the new nine-valent HPV vaccine.
Sigma-Aldrich
Anti-Digoxin–Alkaline Phosphatase antibody, Mouse monoclonal, clone DI-22, purified from hybridoma cell culture