MilliporeSigma
  • Polyethyleneimine/poly-(γ-glutamic acid)/poly(lactide-co-glycolide) nanoparticles for loading and releasing antiretroviral drug.
Polyethyleneimine/poly-(γ-glutamic acid)/poly(lactide-co-glycolide) nanoparticles for loading and releasing antiretroviral drug.
Colloids and surfaces. B, Biointerfaces (2011-07-19)
Yung-Chih Kuo, Hsin-Wei Yu
ABSTRACT
Nanoparticles (NPs) with ternary components of polyethyleneimine (PEI), poly-(γ-glutamic acid) (γ-PGA), and poly(lactide-co-glycolide) (PLGA) were applied to carry and release saquinavir (SQV). Hydrophobic SQV was encapsulated in the particle core composed of PLGA to form SQV-PLGA NPs, and the surface of SQV-PLGA NPs was grafted successively with hydrophilic γ-PGA and PEI (PEI/γ-PGA/SQV-PLGA NPs). The morphological images revealed that PEI/γ-PGA/SQV-PLGA NPs were spheroid-like, in general. An increase in the concentration of didecyl dimethylammonium bromide and a reduction in the dose of SQV enhanced the entrapment efficiency of SQV in PLGA NPs. In addition, an increment in the molecular weight of γ-PGA reduced the grafting efficiency of PEI on γ-PGA/SQV-PLGA NPs. An increase in the weight percentage of PEI enhanced the average particle diameter. However, the grafting efficiency of PEI on γ-PGA/SQV-PLGA NPs and the dissolution rate of SQV from PEI/γ-PGA/SQV-PLGA NPs reduced when the weight percentage of PEI increased. PEI/γ-PGA/SQV-PLGA NPs are an innovative drug delivery system and can be used for antiretroviral trials.
Sigma-Aldrich
Poly(D,L-lactic-co-glycolic acid)-block-poly(glutamic acid), PLGA average Mn 25,000, mol wt (PGlu average Mn 5,000), lactide:glycolide 50:50