MilliporeSigma
  • TRIM5α associates with proteasomal subunits in cells while in complex with HIV-1 virions.
TRIM5α associates with proteasomal subunits in cells while in complex with HIV-1 virions.
Retrovirology (2011-11-15)
Zana Lukic, Stéphane Hausmann, Sarah Sebastian, Justin Rucci, Jaya Sastri, Seth L Robia, Jeremy Luban, Edward M Campbell
ABSTRACT
The TRIM5 proteins are cellular restriction factors that prevent retroviral infection in a species-specific manner. Multiple experiments indicate that restriction activity requires accessory host factors, including E2-enzymes. To better understand the mechanism of restriction, we conducted yeast-two hybrid screens to identify proteins that bind to two TRIM5 orthologues. The only cDNAs that scored on repeat testing with both TRIM5 orthologues were the proteasome subunit PSMC2 and ubiquitin. Using co-immunoprecipitation assays, we demonstrated an interaction between TRIM5α and PSMC2, as well as numerous other proteasome subunits. Fluorescence microscopy revealed co-localization of proteasomes and TRIM5α cytoplasmic bodies. Forster resonance energy transfer (FRET) analysis indicated that the interaction between TRIM5 and PSMC2 was direct. Previous imaging experiments demonstrated that, when cells are challenged with fluorescently-labeled HIV-1 virions, restrictive TRIM5α orthologues assemble cytoplasmic bodies around incoming virion particles. Following virus challenge, we observed localization of proteasome subunits to rhTRIM5α cytoplasmic bodies that contained fluorescently labeled HIV-1 virions. Taken together, the results presented here suggest that localization of the proteasome to TRIM5α cytoplasmic bodies makes an important contribution to TRIM5α-mediated restriction.
Sigma-Aldrich
Anti-PSMC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution