MilliporeSigma
  • Nanoparticles of Titanium and Zinc Oxides as Novel Agents in Tumor Treatment: a Review.
Nanoparticles of Titanium and Zinc Oxides as Novel Agents in Tumor Treatment: a Review.
Nanoscale research letters (2017-03-30)
Janusz Bogdan, Joanna Pławińska-Czarnak, Joanna Zarzyńska
ABSTRACT
Cancer has become a global problem. On all continents, a great number of people are diagnosed with this disease. In spite of the progress in medical care, cancer still ends fatal for a great number of the ill, either as a result of a late diagnosis or due to inefficiency of therapies. The majority of the tumors are resistant to drugs. Thus, the search for new, more effective therapy methods continues. Recently, nanotechnology has been attributed with big expectations in respect of the cancer fight. That interdisciplinary field of science creates nanomaterials (NMs) and nanoparticles (NPs) that can be applied, e.g., in nanomedicine. NMs and NPs are perceived as very promising in cancer therapy since they can perform as drug carriers, as well as photo- or sonosensitizers (compounds that generate the formation of reactive oxygen species as a result of either electromagnetic radiation excitation with an adequate wavelength or ultrasound activation, respectively). Consequently, two new treatment modalities, the photodynamic therapy (PDT) and the sonodynamic therapy (SDT) have been created. The attachment of ligands or antibodies to NMs or to NPs improve their selective distribution into the targeted organ or cell; hence, the therapy effectiveness can be improved. An important advantage of the targeted tumor treatment is lowering the cyto- and genotoxicity of active substance towards healthy cells. Therefore, both PDT and SDT constitute a valuable alternative to chemo- or radiotherapy. The vital role in cancer eradication is attributed to two inorganic sensitizers in their nanosized scale: titanium dioxide and zinc oxide.
Sigma-Aldrich
Titania nanoparticles, dispersion, 5 wt. % in ethanol, 500 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 5 wt. % in ethanol, 900 nm particle size
Sigma-Aldrich
Titania nanoparticles, 300 nm particle size, dispersion, 10 wt. % in ethanol
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in water, 150 nm particle size
Sigma-Aldrich
Titania nanoparticles, 150 nm particle size, dispersion, 5 wt. % in water
Sigma-Aldrich
Titania nanoparticles, dispersion, 5 wt. % in ethanol, 300 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 5 wt. % in water, 500 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 5 wt. % in water, 700 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 5 wt. % in ethanol, 700 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in ethanol, 700 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in ethanol, 500 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in water, 300 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in ethanol, 900 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in water, 500 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in water, 900 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in water, 700 nm particle size
Sigma-Aldrich
Titania nanoparticles, dispersion, 10 wt. % in ethanol, 150 nm particle size
Sigma-Aldrich
Titania nanoparticles, 900 nm particle size, dispersion, 5 wt. % in water
Sigma-Aldrich
Titania nanoparticles, 300 nm particle size, dispersion, 5 wt. % in water