MilliporeSigma
  • Light Regulation of Enzyme Allostery through Photo-responsive Unnatural Amino Acids.
Light Regulation of Enzyme Allostery through Photo-responsive Unnatural Amino Acids.
Cell chemical biology (2019-09-10)
Andrea C Kneuttinger, Kristina Straub, Philipp Bittner, Nadja A Simeth, Astrid Bruckmann, Florian Busch, Chitra Rajendran, Enrico Hupfeld, Vicki H Wysocki, Dominik Horinek, Burkhard König, Rainer Merkl, Reinhard Sterner
ABSTRACT
Imidazole glycerol phosphate synthase (ImGPS) is an allosteric bienzyme complex in which substrate binding to the synthase subunit HisF stimulates the glutaminase subunit HisH. To control this stimulation with light, we have incorporated the photo-responsive unnatural amino acids phenylalanine-4'-azobenzene (AzoF), o-nitropiperonyl-O-tyrosine (NPY), and methyl-o-nitropiperonyllysine (mNPK) at strategic positions of HisF. The light-mediated isomerization of AzoF at position 55 (fS55AzoFE ↔ fS55AzoFZ) resulted in a reversible 10-fold regulation of HisH activity. The light-mediated decaging of NPY at position 39 (fY39NPY → fY39) and of mNPK at position 99 (fK99mNPK → fK99) led to a 4- to 6-fold increase of HisH activity. Molecular dynamics simulations explained how the unnatural amino acids interfere with the allosteric machinery of ImGPS and revealed additional aspects of HisH stimulation in wild-type ImGPS. Our findings show that unnatural amino acids can be used as a powerful tool for the spatiotemporal control of a central metabolic enzyme complex by light.
Sigma-Aldrich
Methyl-o-nitropiperonyllysine, ≥95%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Phenylalanine-4′-azobenzene HCl, ≥95%
Sigma-Aldrich
L-Glutamate Oxidase from Streptomyces sp., recombinant, expressed in E. coli