MilliporeSigma
  • A Small-Molecule Pan-Id Antagonist Inhibits Pathologic Ocular Neovascularization.
A Small-Molecule Pan-Id Antagonist Inhibits Pathologic Ocular Neovascularization.
Cell reports (2019-10-03)
Paulina M Wojnarowicz, Raquel Lima E Silva, Masayuki Ohnaka, Sang Bae Lee, Yvette Chin, Anita Kulukian, Sung-Hee Chang, Bina Desai, Marta Garcia Escolano, Riddhi Shah, Marta Garcia-Cao, Sijia Xu, Rashmi Kadam, Yehuda Goldgur, Meredith A Miller, Ouathek Ouerfelli, Guangli Yang, Tsutomu Arakawa, Steven K Albanese, William A Garland, Glenn Stoller, Jaideep Chaudhary, Larry Norton, Rajesh Kumar Soni, John Philip, Ronald C Hendrickson, Antonio Iavarone, Andrew J Dannenberg, John D Chodera, Nikola Pavletich, Anna Lasorella, Peter A Campochiaro, Robert Benezra
ABSTRACT
Id helix-loop-helix (HLH) proteins (Id1-4) bind E protein bHLH transcription factors, preventing them from forming active transcription complexes that drive changes in cell states. Id proteins are primarily expressed during development to inhibit differentiation, but they become re-expressed in adult tissues in diseases of the vasculature and cancer. We show that the genetic loss of Id1/Id3 reduces ocular neovascularization in mouse models of wet age-related macular degeneration (AMD) and retinopathy of prematurity (ROP). An in silico screen identifies AGX51, a small-molecule Id antagonist. AGX51 inhibits the Id1-E47 interaction, leading to ubiquitin-mediated degradation of Ids, cell growth arrest, and reduced viability. AGX51 is well-tolerated in mice and phenocopies the genetic loss of Id expression in AMD and ROP models by inhibiting retinal neovascularization. Thus, AGX51 is a first-in-class compound that antagonizes an interaction formerly considered undruggable and that may have utility in the management of multiple diseases.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
AGX51, ≥98% (HPLC)
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent