MilliporeSigma
  • Effect of Divalent Cation on Swelling Behavior of Anionic Microgels: Quantification and Dynamics of Ion Uptake and Release.
Effect of Divalent Cation on Swelling Behavior of Anionic Microgels: Quantification and Dynamics of Ion Uptake and Release.
Langmuir : the ACS journal of surfaces and colloids (2019-10-05)
Vittoria Chimisso, Csaba Fodor, Wolfgang Meier
ABSTRACT
Poly(N-vinylcaprolactam-co-itaconate) (P(VCL-co-IADME) microgels were synthesized varying the molar ratio between VCL and IADME via free radical precipitation polymerization in the presence of quaternary ammonium surfactant. In order to determine the effect of the divalent metal ions on the structure and the swelling behavior of the microgel systems, both neutral and charged forms of the hydrogels after hydrolysis were investigated. The triggered gel collapse caused by the divalent metal ion together with the quantification of the metal ion uptake was studied in detail by titration and ion chromatography methods and revealed the minimum concentration around 0.1 mM to trigger gel collapse on the treated gels. Uptake and release dynamics of the gels were followed by turbidity measurements and were in the time-range of 2 and 17 s, depending on the composition and the concentrations.
Sigma-Aldrich
N-Vinylcaprolactam, 98%, stabilized
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
Sigma-Aldrich
Sodium phosphotungstate hydrate, ≥99.9% trace metals basis
Sigma-Aldrich
Dimethyl itaconate, 99%