MilliporeSigma
  • Tunable Fluorescence-Responsive Double Hydrophilic Block Polymers Induced by the Formation of Pseudopolyrotaxanes with Cucurbit[7]Uril.
Tunable Fluorescence-Responsive Double Hydrophilic Block Polymers Induced by the Formation of Pseudopolyrotaxanes with Cucurbit[7]Uril.
Polymers (2019-09-12)
Xiumin Qiu, Xin Wang, Shengzhen Hou, Jin Zhang, Jing Zhou, Yebang Tan
ABSTRACT
There is an urgent need for new strategies that allow the simultaneous detection and control of drug delivery. By making use of supramolecular host-guest interactions, a kind of pseudopolyrotaxanes, as a visualizable nanoscale drug carrier has been constructed by self-assembly of cucurbit[7]uril (CB[7]) with methoxy poly(ethylene glycol)-block-quaternized poly(4-vinyl pyridine) (mPEG-b-QP4VP) using 4-(chloromethyl)benzonitrile. Simple addition of CB[7] into an aqueous solution of mPEG-b-QP4VP resulted in noncovalent attachment of CB[7] to 4-cyanobenzyl-containing polymers, transforming the nonemissive mPEG-b-QP4VP micelles into highly fluorescent micelles. These pseudopolyrotaxanes micelles exhibited remarkable supramolecular assembly-induced emission enhancement and excellent biocompatibility, showing great potential for bioimaging applications. In addition, the efficient cellular uptake of the developed pseudopolyrotaxanes micelles loaded with the anticancer drug doxorubicin was a promising platform for simultaneous cell imaging and drug delivery, thereby widening their application in cancer theranostics.
Sigma-Aldrich
Tris[2-(dimethylamino)ethyl]amine, 97%
Sigma-Aldrich
4-(Chloromethyl)benzonitrile, ≥97.0%
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%