MilliporeSigma
Hybrid Uranyl-Phosphonate Coordination Nanocage.
Inorganic chemistry (2019-09-13)
Pius O Adelani, Josemaria S Soriano, Bryan E Galeas, Ginger E Sigmon, Jennifer E S Szymanowski, Peter C Burns
ABSTRACT
We report herein a general synthetic approach for designing uranyl coordination cages. Compounds 1 and 2 are constructed through a temperature-dependent and solvent-driven self-assembly. In both cases, the synthetic strategy involves in situ phosphonate ligand condensation into a flexible pyrophosphonate ligand. This pyrophosphonate ligand formation is essential for the introduction of curvature into these compounds. In the presence of PF6- ions that are derived from hydrofluoric acid, a macrocyclic uranyl-phosphonate discrete compound, 1, whose cavity contains PF6- ions, hydronium ions, and water molecules, is obtained. When Cs+ cations are used in the synthesis, a remarkable uranyl coordination nanocage, 2, resulted. The macrocycle (1) is approximately 10.9 × 10.9 Å2 in diameter while the nanocage (2) is approximately 15.0 × 11.3 Å2 in diameter, as measured from the outer oxygen atoms of the uranyl centers. Both compounds are constructed from a UO22+ moiety, coordinated by an additional four oxygen atoms from the phosphonate group to form pentagonal bipyramidal geometry. All the compounds fluoresce at room temperature, showing characteristic vibronically coupled charge-transfer based emission.
Sigma-Aldrich
tert-Butylphosphonic acid, 98%