MilliporeSigma
  • SIRT1-regulated HMGB1 release is partially involved in TLR4 signal transduction: A possible anti-neuroinflammatory mechanism of resveratrol in neonatal hypoxic-ischemic brain injury.
SIRT1-regulated HMGB1 release is partially involved in TLR4 signal transduction: A possible anti-neuroinflammatory mechanism of resveratrol in neonatal hypoxic-ischemic brain injury.
International immunopharmacology (2019-07-31)
Kai Le, Enkhmurun Chibaatar Daliv, Shanshan Wu, Fangyuan Qian, Abdoulaye Idriss Ali, Dafan Yu, Yijing Guo
ABSTRACT
Neonatal hypoxic-ischemic brain injury (HIBI) is a knotty disease that lacks appropriate treatment. Inflammation is an important contributor to brain damage, and microglia are responsible for eliciting early and pronounced inflammatory reactions in the immature brain after hypoxic-ischemic (HI) insult. Acetylated HMGB1 can be released from immune cells into the extracellular space, where it acts as a danger-associated molecular pattern molecule to activate TLR4 signalling-mediated inflammatory responses. Resveratrol has neuroprotective and anti-inflammatory effects against HIBI, but whether these effects involve the regulation of the TLR4 signalling pathway and whether HMGB1 participates in this process is still unclear. We investigated the anti-inflammatory effects of resveratrol in HIBI and the molecular mechanisms potentially involved in the effect. The in vivo and in vitro results indicated that the level of cytoplasmic HMGB1 in microglia increased after insult and that treating experimental animals or mouse BV2 microglial cells with resveratrol attenuated HI insult-induced neuroinflammation, which was characterized by improved behavioural defects, reduced microglial activation and TLR4/MyD88/NF-κB signalling, and attenuated primary neuronal damage; this was accompanied by the inhibition of HMGB1 nucleoplasmic transfer and extracellular release. EX527 pretreatment reversed these effects. In addition, co-immunoprecipitation confirmed that SIRT1 was directly involved in the HMGB1 acetylation process in BV2 cells after oxygen glucose deprivation. These data demonstrate that resveratrol plays a neuroprotective role in neonatal HIBI by activating SIRT1 to inhibit HMGB1/TLR4/MyD88/NF-κB signalling and subsequent neuroinflammatory responses.
Sigma-Aldrich
SIRT1 Assay Kit, sufficient for 100 assays