MilliporeSigma
  • Maelstrom Represses Canonical Polymerase II Transcription within Bi-directional piRNA Clusters in Drosophila melanogaster.
Maelstrom Represses Canonical Polymerase II Transcription within Bi-directional piRNA Clusters in Drosophila melanogaster.
Molecular cell (2018-12-12)
Timothy H Chang, Eugenio Mattei, Ildar Gainetdinov, Cansu Colpan, Zhiping Weng, Phillip D Zamore
ABSTRACT
In Drosophila, 23-30 nt long PIWI-interacting RNAs (piRNAs) direct the protein Piwi to silence germline transposon transcription. Most germline piRNAs derive from dual-strand piRNA clusters, heterochromatic transposon graveyards that are transcribed from both genomic strands. These piRNA sources are marked by the heterochromatin protein 1 homolog Rhino (Rhi), which facilitates their promoter-independent transcription, suppresses splicing, and inhibits transcriptional termination. Here, we report that the protein Maelstrom (Mael) represses canonical, promoter-dependent transcription in dual-strand clusters, allowing Rhi to initiate piRNA precursor transcription. Mael also represses promoter-dependent transcription at sites outside clusters. At some loci, Mael repression requires the piRNA pathway, while at others, piRNAs play no role. We propose that by repressing canonical transcription of individual transposon mRNAs, Mael helps Rhi drive non-canonical transcription of piRNA precursors without generating mRNAs encoding transposon proteins.
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), highly cross-adsorbed, CF 594 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Bestatin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
AEBSF, Hydrochloride, AEBSF, CAS 30827-99-7, is a specific irreversible inhibitor of chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. A nontoxic alternative to PMSF and DFP.