MilliporeSigma
  • No Sex-Specific Differences in the Acute Antidepressant Actions of (R)-Ketamine in an Inflammation Model.
No Sex-Specific Differences in the Acute Antidepressant Actions of (R)-Ketamine in an Inflammation Model.
The international journal of neuropsychopharmacology (2018-06-13)
Lijia Chang, Hidetoh Toki, Youge Qu, Yuko Fujita, Akiko Mizuno-Yasuhira, Jun-Ichi Yamaguchi, Shigeyuki Chaki, Kenji Hashimoto
ABSTRACT
Although previous reports suggest sex-specific differences in the antidepressant actions of (R,S)-ketamine, these differences in the antidepressant actions of (R)-ketamine, which is more potent than (S)-ketamine, are unknown. Saline or (R)-ketamine was administered 23 hours post lipopolysaccharide administration to adult male or female mice. Subsequently, antidepressant effects were assessed using a forced swimming test. Furthermore, the concentration of (R)-ketamine and its 2 major metabolites, (R)-norketamine and (2R,6R)-hydroxynorketamine, was measured in the plasma and brain after the administration of (R)-ketamine in the mice. (R)-ketamine (10 mg/kg) significantly attenuated the increased immobility time of forced swimming test in the lipopolysaccharide-treated mice. There were no sex-specific differences in the concentrations of (R)-ketamine and its 2 metabolites in the plasma and brain. These findings showed no sex-specific differences in terms of the acute antidepressant effects and pharmacokinetic profile of (R)-ketamine.