MilliporeSigma
  • Application of the automated haematology analyzer XN-30 for discovery and development of anti-malarial drugs.
Application of the automated haematology analyzer XN-30 for discovery and development of anti-malarial drugs.
Malaria journal (2019-01-16)
Takahiro Tougan, Yuji Toya, Kinya Uchihashi, Toshihiro Horii
ABSTRACT
The erythrocytic stage of Plasmodium falciparum parasites in humans is clinically important, as the parasites at this growth stage causes malarial symptoms. Most of the currently available anti-malarial drugs target this stage, but the emergence and spread of parasites resistant to anti-malarial drugs are a major challenge to global eradication efforts; therefore, the development of novel medicines is urgently required. In this study, the in vitro anti-malarial activity of five current anti-malarial drugs (artemisinin, atovaquone, chloroquine, mefloquine, and pyrimethamine) and 400 compounds from the Pathogen Box provided by the Medicines for Malaria Venture on P. falciparum parasites was characterized using the XN-30 analyzer. Furthermore, the outcomes obtained using the analyser were classified according to the parasitaemias of total and each developmental stages. The growth inhibition rate and the half-maximal (50%) inhibitory concentration (IC50) of the five current anti-malarial drugs were calculated from the parasitaemia detected using the XN-30 analyzer. Respective strains and drugs presented strongly fitted sigmoidal curves, and the median SD at all tested concentrations was 1.6, suggesting that the variation in values measured with the analyser was acceptably low for the comparison of drug efficacy. Furthermore, the anti-malarial activity of the 400 compounds from the Pathogen Box was tested, and 141 drugs were found to be effective. In addition, the efficacy was classified into 4 types (Type I, parasites were arrested or killed without DNA replication; Type II, parasites were arrested or killed similar to Type I, and the parasitaemia was apparently decreased; Type III, parasites progressed to trophozoite without sufficient DNA replication; and Type IV, parasites were arrested at late trophozoite or schizont after DNA replication). The current study demonstrates that the XN-30 analyzer objectively, reproducibly, and easily evaluated and characterized the anti-malarial efficacy of various compounds. The results indicate the potential of the XN-30 analyzer as a powerful tool for drug discovery and development in addition to its use as an important diagnostic tool.