MilliporeSigma
  • IL-6 signaling contributes to radioresistance of prostate cancer through key DNA repair-associated molecules ATM, ATR, and BRCA 1/2.
IL-6 signaling contributes to radioresistance of prostate cancer through key DNA repair-associated molecules ATM, ATR, and BRCA 1/2.
Journal of cancer research and clinical oncology (2019-04-26)
Xiaodong Chen, Feng Chen, Yu Ren, Guobin Weng, Lijun Xu, Xiang Xue, Peter C Keng, Soo Ok Lee, Yuhchyau Chen
ABSTRACT
To study an association between IL-6 signaling and resistance to radiotherapy of prostate cancer (PCa) and explore the molecular mechanisms involved. IL-6 expressing C4-2 and CWR22Rv1 (C4-2IL-6/CWRIL-6) and vector control (C4-2vec/CWRvec) cell lines were developed. Radiation-sensitivities of these cells were compared in clonogenic assay, Comet assay, and γH2AX staining. In xenograft animal studies, radiation-sensitivity of C4-2IL-6 cell-derived tumors vs. C4-2vec cell-derived tumors was investigated. To reveal IL-6 downstream molecules involved in DNA repair after radiation, qPCR and Western blot analyses as well as immunofluorescence staining were performed. Transcriptional control of IL-6 on ATM and ATR molecules was also investigated. We found C4-2IL-6 and CWRIL-6 cells survived better than their vector control cells after irradiation, and animal studies confirmed such in vitro results. We discovered that DNA repair-related molecules such as ATM, ATR, BRCA1, and BRCA2 were significantly upregulated in irradiated IL-6 expressing cells compared with vector control cells. We further defined that IL-6 signaling regulated cellular expressions of ATM and ATR at the transcriptional level through the activation of Stat3 signaling pathway. IL-6 leads to PCa resistance to radiation through upregulation of DNA repair associated molecules ATM, ATR, BRCA1, and BRCA2.