MilliporeSigma
Hepatic spheroids used as an in vitro model to study malaria relapse.
Biomaterials (2019-06-14)
Adeline C Y Chua, Abhishek Ananthanarayanan, Jessica Jie Ying Ong, Jen Yi Wong, Andy Yip, Nisha Hari Singh, Yinghua Qu, Laurent Dembele, Michael McMillian, Ratawan Ubalee, Silas Davidson, Anchalee Tungtaeng, Rawiwan Imerbsin, Kapish Gupta, Chiara Andolina, Fan Lee, Kevin S-W Tan, François Nosten, Bruce Russell, Amber Lange, Thierry T Diagana, Laurent Rénia, Bryan K S Yeung, Hanry Yu, Pablo Bifani
ABSTRACT
Hypnozoites are the liver stage non-dividing form of the malaria parasite that are responsible for relapse and acts as a natural reservoir for human malaria Plasmodium vivax and P. ovale as well as a phylogenetically related simian malaria P. cynomolgi. Our understanding of hypnozoite biology remains limited due to the technical challenge of requiring the use of primary hepatocytes and the lack of robust and predictive in vitro models. In this study, we developed a malaria liver stage model using 3D spheroid-cultured primary hepatocytes. The infection of primary hepatocytes in suspension led to increased infectivity of both P. cynomolgi and P. vivax infections. We demonstrated that this hepatic spheroid model was capable of maintaining long term viability, hepatocyte specific functions and cell polarity which enhanced permissiveness and thus, permitting for the complete development of both P. cynomolgi and P. vivax liver stage parasites in the infected spheroids. The model described here was able to capture the full liver stage cycle starting with sporozoites and ending in the release of hepatic merozoites capable of invading simian erythrocytes in vitro. Finally, we showed that this system can be used for compound screening to discriminate between causal prophylactic and cidal antimalarials activity in vitro for relapsing malaria.
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture