MilliporeSigma
Therapeutic Potential of OMe-PS-miR-29b1 for Treating Liver Fibrosis.
Molecular therapy : the journal of the American Society of Gene Therapy (2018-10-06)
Virender Kumar, Vinod Kumar, Jiangtao Luo, Ram I Mahato
ABSTRACT
Trans-differentiation of quiescent hepatic stellate cells (HSCs) into active myofibroblasts secretes excess amounts of extracellular matrix (ECM) proteins. miR-29b1 has the potential to treat liver fibrosis, because it targets several profibrotic genes. We previously demonstrated that miR-29b1 and the hedgehog (Hh) pathway inhibitor GDC-0449 could, together, inhibit the activation of HSCs and ECM production in common bile-duct-ligated (CBDL) mice. Herein, we determined the effect of chemical modifications of miR-29b1 on its stability, immunogenicity, and Argonaute-2 (Ago2) loading in vitro, after modifying its antisense strand with phosphorothioate (PS-miR-29b1), 2'-O-methyl-phosphorothioate (OMe-miR-29b1), locked nucleic acid (LNA-miR-29b1), and N,N'-diethyl-4-(4-nitronaphthalen-1-ylazo)-phenylamine (ZEN-miR-29b1). Chemical modifications significantly improved stability of miR-29b1 in 50% FBS. Among all the modified miRNAs tested, OMe-PS-miR-29b1 showed the highest stability with low immunogenicity, without the loss of efficacy in vitro. Therefore, OMe-PS-miR-29b1 was complexed with poly(ethylene glycol)-block-poly(2-methyl-2-carboxyl-propylenecarbonate-graft-dodecanol-graft-tetraethylenepentamine (mPEG-b-PCC-g-DC-g-TEPA) cationic micelles, and anti-fibrotic efficacy was evaluated in CBDL mice. There was a significant improvement in liver histology and decrease in the levels of injury markers. Further, mRNA/protein levels of collagen, α-SMA, and TIMP-1 were significantly lower for the OMe-PS-miR-29b1-loaded micelles compared to miR-29b1-loaded micelles. In conclusion, micellar delivery of OMe-PS-miR-29b1 is a promising strategy to treat liver fibrosis.
Sigma-Aldrich
Anti-alpha-Actin Antibody, Smooth Muscle, from rabbit, purified by affinity chromatography