MilliporeSigma
  • Differential suppression of delayed-rectifier and inwardly rectifier K+ currents by a group of ent-kaurane-type diterpenoids from Croton tonkinensis, in microglial cells.
Differential suppression of delayed-rectifier and inwardly rectifier K+ currents by a group of ent-kaurane-type diterpenoids from Croton tonkinensis, in microglial cells.
European journal of pharmacology (2019-05-28)
Hung-Tsung Hsiao, Yu-Chi Lee, Yen-Chin Liu, Ping-Chung Kuo, Sheng-Nan Wu
ABSTRACT
Croton is an extensive flowering plant genus in the spurge family, Euphorbiaceae. Three croton compounds with the common ent-kaurane skeleton were purified from Croton tonkinensis. By using patch-clamp recording technique, we thoroughly examined the effect of a group of croton compounds, croton-01 (ent-18-acetoxy-7α-hydroxykaur-16-en-15-one), croton-02 (ent-7α,14β-dihydroxykaur-16-en-15-one), and croton-03 (ent-1β-acetoxy-7α,14β-dihydroxykaur-16-en-15-one), on the membrane current in SM826 and BV2 microglial cells. Although neither voltage-gated Na+ nor Ca2+ currents were present in these cells, both delayed-rectifier K+ outward (IK(DR)) and inwardly rectifying K+ currents (IK(IR)) were readily detected. Croton-03 differentially caused inhibition of IK(DR) or IK(IR) in a concentration-dependent manner. According to a minimal scheme, the shortening of the time constant in either the IK(DR)-related block or IK(IR) caused by different concentrations of croton-03 was quantitatively estimated with a dissociation constant of 6.45 and 29.5 μM, respectively. In SM826 cells differentiated with β-amyloid, inhibitory action on these K+ currents remained unaltered. In ultraviolet C-irradiated cells, the magnitude of IK(IR) was still decreased by addition of croton-03. Therefore, our study suggests that these ent-kaurane diterpenoids ought to somehow act on the cellular mechanisms by which they influence the functional activities of microglial cells.