MilliporeSigma
  • Leptin signaling impairs macrophage defenses against Salmonella Typhimurium.
Leptin signaling impairs macrophage defenses against Salmonella Typhimurium.
Proceedings of the National Academy of Sciences of the United States of America (2019-07-28)
Julia Fischer, Saray Gutièrrez, Raja Ganesan, Chiara Calabrese, Rajeev Ranjan, Gökhan Cildir, Nina Judith Hos, Jan Rybniker, Martina Wolke, Jochen W U Fries, Vinay Tergaonkar, Georg Plum, Adam Antebi, Nirmal Robinson
ABSTRACT
The dynamic interplay between metabolism and immune responses in health and disease, by which different immune cells impact on metabolic processes, are being increasingly appreciated. However, the potential of master regulators of metabolism to control innate immunity are less understood. Here, we studied the cross-talk between leptin signaling and macrophage function in the context of bacterial infections. We found that upon infection with Gram-negative pathogens, such as Salmonella Typhimurium, leptin receptor (Lepr) expression increased in both mouse and human macrophages. Unexpectedly, both genetic Lepr ablation in macrophages and global pharmacologic leptin antagonization augmented lysosomal functions, reduced S Typhimurium burden, and diminished inflammation in vitro and in vivo. Mechanistically, we show that leptin induction activates the mTORC2/Akt pathway and subsequently down-regulates Phlpp1 phosphatase, allowing for phosphorylated Akt to impair lysosomal-mediated pathogen clearance. These data highlight a link between leptin signaling, the mTORC2/Phlpp1/Akt axis, and lysosomal activity in macrophages and have important therapeutic implications for modulating innate immunity to combat Gram-negative bacterial infections.
Sigma-Aldrich
Anti-PHLPP1 Antibody, from rabbit, purified by affinity chromatography