MilliporeSigma
  • Liposome formulations of o-carborane for the boron neutron capture therapy of cancer.
Liposome formulations of o-carborane for the boron neutron capture therapy of cancer.
Biophysical chemistry (2019-02-24)
Temidayo O B Olusanya, Gianpiero Calabrese, Dimitrios G Fatouros, John Tsibouklis, James R Smith
ABSTRACT
Based on the promise of liposomes as convenient vehicles for the transport of boronated agents for the boron neutron capture therapy (BCNT) of cancer, this paper reports a method for the formulation and characterisation of stable o-carborane-loaded liposomes (ca. 80-100 nm) of dipalmitoyl-phosphatidylcholine (DPPC) or 1,2-distearol-sn-glycerol-3-phosphocholine (DSPC). Preliminary pharmaceutical characterisation experiments have demonstrated the integrity of both DPPC and DSPC liposomal membranes in serum and in PBS and also indicate that these o-carborane-loaded liposomes are candidate carrier vehicles for further evaluation with a view to exploitation in BNCT.