MilliporeSigma
  • Reprogramming Probiotic Lactobacillus reuteri as a Biosensor for Staphylococcus aureus Derived AIP-I Detection.
Reprogramming Probiotic Lactobacillus reuteri as a Biosensor for Staphylococcus aureus Derived AIP-I Detection.
ACS synthetic biology (2018-04-14)
David Lubkowicz, Chun Loong Ho, In Young Hwang, Wen Shan Yew, Yung Seng Lee, Matthew Wook Chang
ABSTRACT
Gram-positive Staphylococcus aureus infection that results in pneumonia, urinary tract infection, and in severe cases, sepsis, has recently been classified as a serious threat to public health. Rapid and cost-effective detection of these infections are costly and time-consuming. Here, we present probiotic lactic acid bacteria engineered to detect autoinducer peptide-I (AIP-I), a quorum sensing molecule produced by Staphylococcus sp. during pathogenesis. We achieved this by adapting the well-characterized agr quorum sensing ( agrQS) from Staphylococcus aureus into Lactobacillus reuteri. The engineered biosensor is able to detect AIP-I levels in the nanomolar to micromolar range. We further investigated the function of the biosensor to detect real-time changes in AIP-I levels to understand the dynamics of Staphylococcus aureus under various strenuous conditions. The developed sensors would be useful for detection of Staphylococcus contamination in hospital settings and for high-throughput drug screening.