MilliporeSigma
  • Lenalidomide Enhances the Function of CS1 Chimeric Antigen Receptor-Redirected T Cells Against Multiple Myeloma.
Lenalidomide Enhances the Function of CS1 Chimeric Antigen Receptor-Redirected T Cells Against Multiple Myeloma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2017-10-25)
Xiuli Wang, Miriam Walter, Ryan Urak, Lihong Weng, Christian Huynh, Laura Lim, ChingLam W Wong, Wen-Chung Chang, Sandra H Thomas, James F Sanchez, Lu Yang, Christine E Brown, Flavia Pichiorri, Myo Htut, Amrita Y Krishnan, Stephen J Forman
ABSTRACT
Purpose: Multiple myeloma remains an incurable malignancy of plasma cells despite considerable advances in treatment. The purpose of the study was to develop novel chimeric antigen receptors (CAR) for the treatment of multiple myeloma and explore combinatorial therapy using CAR T cells and immunomodulatory drugs such as lenalidomide for increasing treatment efficacy.Experimental Design: We redirected central memory T cells to express second-generation CAR-specific for CS1 and adoptively transferred them into multiple myeloma tumor-bearing mice to test their anti-multiple myeloma activity. CS1 CAR T cells were transduced and expanded in the presence of lenalidomide in vitro The phenotype and effector function of CS1 CAR T cells treated with and without lenalidomide were compared. Finally, CS1 CAR T cells and lenalidomide were administered to treat multiple myeloma-bearing mice as combinatorial therapy.Results: CS1 CAR T cells exhibited efficient antitumor activity when adoptively transferred into mice. Mechanistic studies indicated that the addition of lenalidomide during CS1 CAR T-cell expansion in vitro enhanced the immune functions of CS1 CAR T cells, including cytotoxicity, memory maintenance, Th1 cytokine production, and immune synapse formation. Furthermore, lenalidomide enhanced the antitumor activity and persistence of adoptively transferred CS1 CAR T cells in vivoConclusions: The study demonstrates that lenalidomide improves the anti-multiple myeloma properties of CS1-directed CAR T cells and provides a basis for a planned clinical trial using the combination of lenalidomide with engineered T cells against CS1 in relapsed myeloma. Clin Cancer Res; 24(1); 106-19. ©2017 AACR.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution