MilliporeSigma
  • Chloroplast-to-Nucleus Signaling Regulates MicroRNA Biogenesis in Arabidopsis.
Chloroplast-to-Nucleus Signaling Regulates MicroRNA Biogenesis in Arabidopsis.
Developmental cell (2019-01-01)
Xiaofeng Fang, Gaozhan Zhao, Su Zhang, Yaoxi Li, Hanqing Gu, Yan Li, Qiao Zhao, Yijun Qi
ABSTRACT
As integral regulators in plant development and stress response, microRNAs (miRNAs) themselves need to be tightly regulated. Here, we show that tocopherols (vitamin E), lipid-soluble antioxidants synthesized from tyrosine in chloroplasts, positively regulate the biogenesis of miRNAs. Tocopherols are required for the accumulation of 3'-phosphoadenosine 5'-phosphate (PAP), a retrograde inhibitor of the nuclear exoribonucleases (XRN), which may protect primary miRNAs from being degraded and promote mature miRNA production. Such regulation is involved in heat-induced accumulation of miR398 and plant acquisition of heat tolerance. Our study reveals a chloroplast-to-nucleus signaling mechanism that favors miRNA biogenesis under heat and possibly other environmental perturbations.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%