MilliporeSigma
  • Phenotypic plasticity of mesenchymal stem cells is crucial for mesangial repair in a model of immunoglobulin light chain-associated mesangial damage.
Phenotypic plasticity of mesenchymal stem cells is crucial for mesangial repair in a model of immunoglobulin light chain-associated mesangial damage.
Ultrastructural pathology (2018-04-19)
Guillermo A Herrera, Jiamin Teng, Chun Zeng, Hongzhi Xu, Man Liang, J Steven Alexander, Bing Liu, Chris Boyer, Elba A Turbat-Herrera
ABSTRACT
Mesangiopathies produced by glomerulopathic monoclonal immunoglobulin light chains (GLCs) acting on the glomerular mesangium produce two characteristic lesions: AL-amyloidosis (AL-Am) and light chain deposition disease (LCDD). In both cases, the pathology is centered in the mesangium, where initial and progressive damage occurs. In AL-Am the mesangial matrix is destroyed and replaced by amyloid fibrils and in LCDD, the mesangial matrix is increased and remodeled. The collagen IV rich matrix is replaced by tenascin. In both conditions, mesangial cells (MCs) become apoptotic as a direct effect of the GLCs. MCs were incubated in-vitro with GLCs and animal kidneys were perfused ex-vivo via the renal artery with GLCs, producing expected lesions, and then mesenchymal stem cells (MSCs) were added to both platforms. Each of the two platforms provided unique information that when put together created a comprehensive evaluation of the processes involved. A "cocktail" with growth and differentiating factors was used to study its effect on mesangial repair. MSCs displayed remarkable phenotypic plasticity during the repair process. The first role of the MSCs after migrating to the affected areas was to dispose of the amyloid fibrils (in AL-Am), the altered mesangial matrix (in LCDD) and apoptotic MCs/debris. To accomplish this task, MSCs transformed into facultative macrophages acquiring an abundance of lysosomes and endocytotic capabilities required to engage in phagocytic functions. Once the mesangial cleaning was completed, MSCs transformed into functional MCs restoring the mesangium to normal. "Cocktail" made the repair process more efficient.