MilliporeSigma
  • Multivalent II [beta-D-Galp-(1-->4)-beta-D-GlcpNAc] and Talpha [beta-D-Galp-(1-->3)-alpha-D-GalpNAc] specific Moraceae family plant lectin from the seeds of Ficus bengalensis fruits.
Multivalent II [beta-D-Galp-(1-->4)-beta-D-GlcpNAc] and Talpha [beta-D-Galp-(1-->3)-alpha-D-GalpNAc] specific Moraceae family plant lectin from the seeds of Ficus bengalensis fruits.
Carbohydrate research (2007-03-16)
Biswajit Singha, Mausumi Adhya, Bishnu P Chatterjee
ABSTRACT
A galactose specific lectin was isolated from the seeds of Ficus bengalensis (Moraceae) fruits and designated as F. bengalensis agglutinin (FBA). The lectin was purified by affinity repulsion chromatography on fetuin-agarose and was a monomer of molecular mass 33kDa. Like other Moraceae family lectins, carbohydrate-binding activity of FBA was independent of any divalent cation. FBA did not bind with simple saccharides, however sugar ligands with aromatic aglycons showed pronounced binding. The combining site of FBA recognized preferably Galbeta1,4GlcNAcbeta1-(II) followed by Galbeta1,3GalNAcalpha1-(Talpha) containing glycotopes. Interaction with saccharides revealed that the combining site of FBA could well accommodate a tetrasaccharide, asialo GM1 glycan (Galbeta1,3GalNAcbeta1,4Galbeta1,4Glcbeta1-), whereas polyvalent Tn (GalNAcalpha1-Ser/Thr), one of the well-recognized ligands of Moraceae family lectin, did not interact well with FBA.
Sigma-Aldrich
Fetuin-Agarose, saline suspension