MilliporeSigma
  • RNA degradosomes exist in vivo in Escherichia coli as multicomponent complexes associated with the cytoplasmic membrane via the N-terminal region of ribonuclease E.
RNA degradosomes exist in vivo in Escherichia coli as multicomponent complexes associated with the cytoplasmic membrane via the N-terminal region of ribonuclease E.
Proceedings of the National Academy of Sciences of the United States of America (2001-01-03)
G G Liou, W N Jane, S N Cohen, N S Lin, S Lin-Chao
ABSTRACT
RNase E isolated from Escherichia coli is contained in a multicomponent "degradosome" complex with other proteins implicated in RNA decay. Earlier work has shown that the C-terminal region of RNase E is a scaffold for the binding of degradosome components and has identified specific RNase E segments necessary for its interaction with polynucleotide phosphorylase (PNPase), RhlB RNA helicase, and enolase. Here, we report electron microscopy studies that use immunogold labeling and freeze-fracture methods to show that degradosomes exist in vivo in E. coli as multicomponent structures that associate with the cytoplasmic membrane via the N-terminal region of RNase E. Whereas PNPase and enolase are present in E. coli in large excess relative to RNase E and therefore are detected in cells largely as molecules unlinked to the RNase E scaffold, immunogold labeling and biochemical analyses show that helicase is present in approximately equimolar amounts to RNase E at all cell growth stages. Our findings, which establish the existence and cellular location of RNase E-based degradosomes in vivo in E. coli, also suggest that RNA processing and decay may occur at specific sites within cells.
Sigma-Aldrich
Polynucleotide phosphorylase from Synechocystis sp., recombinant, expressed in E. coli