MilliporeSigma
  • The role of calcium and N-linked glycans in the oligomerization and carbohydrate binding properties of human immunodeficiency virus external envelope glycoprotein.
The role of calcium and N-linked glycans in the oligomerization and carbohydrate binding properties of human immunodeficiency virus external envelope glycoprotein.
Glycoconjugate journal (1994-04-01)
M Haidar, N Seddiki, J C Gluckman, L Gattegno
ABSTRACT
Envelope glycoproteins of human immunodeficiency virus (gp120 and gp41) occur as oligomers. Here, we show by gel filtration analysis that gp120 oligomerization in vitro is calcium- and temperature-dependent. Recombinant gp120 (rgp120) species were recovered as monomers at 20 degrees C in the absence of calcium, but as tetramers at 37 degrees C in 10 mM CaCl2. Under the latter condition, N-glycanase-deglycosylated rgp120 formed hexamers. Relative to intact rgp120, which has been reported to display carbohydrate-binding properties for N-acetyl-beta-D-glucosaminyl and mannosyl residues, deglycosylation enhanced rgp120 specific binding to mannose-divinylsulfone-agarose, para-aminophenyl-beta-D-GlcNAc-agarose and fetuin-agarose matrices. Taken together, these results rule out the role of homologous lectin-carbohydrate interactions via N-linked glycans in the rgp120 oligomerization, even though its lectin properties may also be calcium-dependent. Deglycosylation may unmask domains of rgp120 polypeptide backbone that independently play a role either in rgp120 lectin activity or in calcium-dependent oligomerization.
Sigma-Aldrich
Fetuin-Agarose, saline suspension