MilliporeSigma
  • A component of BRAF-HDAC complex, BHC80, is required for neonatal survival in mice.
A component of BRAF-HDAC complex, BHC80, is required for neonatal survival in mice.
FEBS letters (2006-05-11)
Shigeki Iwase, Naomi Shono, Arata Honda, Tomoko Nakanishi, Shin-ichi Kashiwabara, Satoru Takahashi, Tadashi Baba
ABSTRACT
BHC80 is a component of BRAF-HDAC complex (BHC) involved in transcriptional repression of neuron-specific genes in non-neuronal cells. However, BHC80 is present in both neuronal and non-neuronal cells. To explore the physiological importance of BHC80 in vivo, and the precise mechanism underlying neuron-specific gene repression by BHC80, we have produced mutant mice lacking Bhc80. The loss of Bhc80 resulted in neonatal lethality without sucking mother's breast milk sufficiently. Although Bhc80-deficient mice showed no developmental defect in the neuronal and non-neuronal tissues, Bhc80 is indispensable for the survival of neonatal pups.
Sigma-Aldrich
Acetyl-Histone H4 Antibody Set, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit